Learn More
Day Impex™ LabGUARD Antimicrobial Hand Soap
Product Code. 12308667
Change view
Click to view available options
Volume (Metric):
1 L
5 L
Unit Size:
Pack of 2
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12308667 5 L Pack of 2
12398657 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12308667 Supplier Day Impex™ Supplier No. 331003
Gently but effectively destroy a broad spectrum of organisms. Day Impex™ LabGUARD Antimicrobial Hand Soap is a concentrated coconut based lotion, specially formulated for frequent or repeated use without drying the skin.
Concentrated
Coconut-based lotion
Safe to use as a shower soap
Natural moisturisers and conditioners
Combats the effects of water, harsh solutions and other irritants found in the laboratory
Specifications
Product Type Hand Soap
Composition Coconut Based Lotion
For Use With (Application) Use as Shower Soap, Combating the Effects of Water, Harsh Solutions and Other Irritants Found in the Laboratory
Quantity 2 Pk.
Volume (Metric) 5 L