Learn More
Macherey Nagel Antibiotic Resistance Test Disks, MN 827 ATD
Used to test resistance of pathogens to antibiotics.
Brand:  Macherey Nagel 484001
Product Code. 10338531
  • 36.20 EUR / Pack of 1000
Only null left
In Stock
Description
  • These products are used in testing the resistance of pathogens to antibiotics
  • For this test the filter paper sections can be impregnated with the antibiotic to be tested and placed on the inoculated nutrient medium
  • Depending on the effectiveness, a smaller or larger zone of inhibition is formed
Specifications
Specifications
Basis weight: 270 g/m2, Filtration speed: 12 sec., Thickness: 0.7 mm
1000/Pk.
9 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions