missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Myb Monoclonal Antibody (IOA3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA190154
This item is not returnable.
View return policy
Description
Predicted molecular weight of ~77 kDa.
Transcriptional activator; DNA-binding protein that specifically recognize the sequence 5'-YAAC[GT]G-3'. Plays an important role in the control of proliferation and differentiation of hematopoietic progenitor cells.
Specifications
| Myb | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P06876, P10242 | |
| MYB | |
| Recombinant v-myb protein, DNA binding protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Frog, Zebrafish | |
| Antibody | |
| IgG2a |
| Immunoprecipitation, Western Blot | |
| IOA3 | |
| Unconjugated | |
| MYB | |
| AI550390; Cmyb; c-myb; cmyb protein; c-myb_CDS; c-myb10A_CDS; c-myb13A_CDS; c-myb14A_CDS; c-myb8B_CDS; efg; gag-myb protein; M16449; MYB; myb proto-oncogene protein; MYB proto-oncogene, transcription factor; Mybl2; myb-related protein B; myeloblastosis oncogene; myeloblastosis oncogene-like 2; myeloblastosis proto-oncogene product; oncogene AMV; proto-oncogene c-Myb; RP1-32B1.1; transcription factor cmyb; transcriptional activator Myb; tumor-specific myb protein; v-myb avian myeloblastosis viral oncogene homolog | |
| Mouse | |
| Protein G | |
| RUO | |
| 17863, 30519, 4602 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction