missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TGR5 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA534262

Product Code. 15365091

  • 522.00 EUR / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey, Marmoset, Rabbit (94%).

This gene encodes a member of the G protein-coupled receptor (GPCR) superfamily. This enzyme functions as a cell surface receptor for bile acids. Treatment of cells expressing this GPCR with bile acids induces the production of intracellular cAMP, activation of a MAP kinase signaling pathway, and internalization of the receptor. The receptor is implicated in the suppression of macrophage functions and regulation of energy homeostasis by bile acids. Alternative splicing results in multiple transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TGR5
Polyclonal
Unconjugated
GPBAR1
BG37; C16DUPq36.3; DUP7q36.3; G protein-coupled bile acid receptor 1; G protein-coupled receptor; G protein-coupled receptor TGR5; Gpbar1; GPCR; GPCR HBG37; GPCR19; GPR131; G-protein coupled bile acid receptor 1; G-protein coupled bile acid receptor BG37; G-protein coupled receptor GPCR19; hBG37; Helodermin-preferring VIP receptor; hGPCR19; M-BAR; membrane bile acid receptor; membrane-type receptor for bile acids; PACAP type III receptor; PACAP-R3; PACAP-R-3; pituitary adenylate cyclase-activating polypeptide type III receptor; Tgr5; vasoactive intestinal peptide receptor 2; vasoactive intestinal polypeptide receptor 2; vasopressive intestinal peptide receptor 2; VIP and PACAP receptor 2; VIP receptor subtype 2; Vip2; VIP2R; VIPR2; VIP-R-2; VPAC2; VPAC2 receptor; VPAC2R; VPCAP2R
Rabbit
Antigen affinity chromatography
RUO
151306
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.87 mg/mL
PBS with 0.1% sodium azide
Q8TDU6
GPBAR1
Synthetic 18 amino acid peptide from 3rd cytoplasmic domain of human TGR5.
50 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.