missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Anti-SARS-CoV-2 Antibody IgM Titer Serologic Assay kit (Spike protein RBD)

Product Code. 17146801
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests

Product Code. 17146801

Brand: ACROBiosystems TASK01396tests

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

kit employs a standard indirect-ELISA format, providing a rapid detection of Anti-SARS-CoV-2 antibodies in serum

The newly identified Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2) has posed a serious threat to human health. A rapid and effective assay kit detecting the levels of anti-SARS-CoV-2 in human serum can facilitate research on characterization of antibodies produced in response to SARS-CoV-2 infection.

Specifications

Product Type Antibody titer assay kits
Kit Contents This assay kit employs a standard capture-ELISA format, providing a rapid detection of anti-SARS-CoV-2 antibody, IgM in serum by SARS-CoV-2 Spike RBD protein. The kit consists of HPR-Spike RBD protein, an Anti-SARS-CoV-2 Antibody (Control, IgM), an anti-
Quantity 96 Tests
Shipping Condition RT
Storage Requirements -20°C, aviod light
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.