missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Anti-SARS-CoV-2 Antibody IgG4 Titer Serologic Assay kit

Product Code. 17186801
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests

Product Code. 17186801

Brand: ACROBiosystems TASK01796tests

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

kit employs a standard indirect-ELISA format, providing a rapid detection of Anti-SARS-CoV-2 antibodies in serum

The ongoing SARS-COV-2 pandemics has sickened 3.7 million people around the world, claiming over 1,000,000 lives. To deal with this global public health crisis, unprecedented efforts are being made to study the virus, SARS-CoV-2. At present, there are 193 new crown vaccine projects registered with the World Health Organization are being developed. To support these studies, ACRO has developed Anti-SARS-CoV-2 subtype antibody Serologic Assay kit including:IgG1, IgG2 IgG3, IgG4.

Specifications

Product Type Antibody titer assay kits
Kit Contents This assay kit employs a standard indirect-ELISA format, providing a rapid detection of Anti-SARS-CoV-2 antibodies in serum by SARS-CoV-2 Spike RBD. The Kit consists of Pre-coated with SARS-CoV-2 Spike RBD Microplate, an Anti-SARS-CoV-2 Antibody (Control)
Quantity 96 Tests
Shipping Condition RT
Storage Requirements 2°C to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.