missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Anti-SARS-CoV-2 Antibody IgG Titer Serologic Assay kit (Spike protein)

Product Code. 17186791
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests

Product Code. 17186791

Brand: ACROBiosystems TASK00796tests

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

kit employs a standard indirect-ELISA format, providing a rapid detection of Anti-SARS-CoV-2 antibodies in serum

The newly identified 2019 novel coronavirus (SARS-CoV-2) has posed a serious threat to human health, and it spreads quickly.This kit is developed for detection of IgG titer of Anti-SARS-CoV-2 antibody in serum/plasma in vitro.

Specifications

Product Type Antibody titer assay kits
Kit Contents This assay kit employs a standard indirect-ELISA format, providing a rapid detection of anti-SARS-CoV-2 IgG in serum by SARS-CoV-2 Spike Protein. The kit consists of High-bind Plate, SARS-CoV-2 Spike Protein, an Anti-SARS-CoV-2 Antibody (Control, IgG), an
Quantity 96 Tests
Shipping Condition RT
Storage Requirements 2°C to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.