Learn More
Invitrogen™ Rotavirus Polyclonal Antibody
Description
PA1-85845 detects human rotavirus serotypes I, II, III, IV, and VIII. Sheep anti Rotavirus antibody recognizes human rotavirus serotypes I, II, III, IV and VIII.
Rotavirus is a double-stranded RNA virus in the family Reoviridae. Five species of rotavirus (A-E) cause disease in animals with species A being the most common in human. Within rotavirus A there are different strains, called serotypes, based on the dual classification system of two proteins on the surface of the virus. Six viral proteins (VPs), or structural proteins, form the virion and six non-structural proteins (NSPs) are expressed in infected cells. Rotavirus is the most common cause of severe diarrhoea among infants and young children. Rotavirus gastroenteritis is a mild to severe disease characterised by vomiting, watery diarrhoea, and low-grade fever, but still leads to hundreds of thousands of deaths each year in developing countries.
Specifications
Specifications
| Antigen | Rotavirus |
| Applications | ELISA, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene Alias | Reoviridae |
| Host Species | Sheep |
| Immunogen | Five serotypes of group A Rotavirus strains Wa (serotype I), DS-1 (serotype II), Ito (III), HOCHI (IV) and 69M (VIII). |
| Purification Method | Protein G |
| Show More |