missing translation for 'onlineSavingsMsg'
Learn More

RIP with control Mouse anti-Human, Unlabeled, Clone: G322-2, BD

Product Code. 15836488
Change view
Click to view available options
Quantity:
150 μg
50 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15836488 150 μg Each
15826488 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15836488 Supplier BD Biosciences Supplier No. 551042

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RIP (receptor interacting protein) is a 74 kDa serine/threonine kinase which may be recruited to TNFR type 1 and Fas (CD95) receptor signal complexes following ligand binding. RIP interacts with other signal proteins within these complexes (e.g., RAIDD) and has also been shown to interact with pro-caspase-2. RIP contains an N-terminal kinase domain as well as a C-terminal death domain that is homologous to intracellular death domain of Fas. Over expression of RIP in vitro is sufficient to induce cell death, demonstrating that RIP functions as an apoptosis-inducing protein. Interaction of the Fas death domain with other intracellular proteins like RIP is an important step leading to downstream components in apoptotic signaling pathways.

Clone G322-2 recognizes human RIP. A recombinant truncated human RIP:tagged fusion protein, lacking the kinase domain of RIP, was used as immunogen. The specificity of the antibody was verified by ELISA, immunoprecipitation and western blot analysis. The antibody is routinely tested by western blot analysis in human Jurkat T cells where it recognizes RIP as a 74 kDa band. Smaller molecular weight breakdown bands of ∽30 kDa, 22 kDa, and/or 16 kDa are sometimes observed.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RIP with control
Applications Western Blot
Classification Monoclonal
Clone G322-2
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Primary or Secondary Primary
Includes Purified Mouse Anti-Human RIP (150μg) 3vials of 50μg, Jurkat Cell Lysate (50μg)
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.