missing translation for 'onlineSavingsMsg'
Learn More
Learn More
polymerase I and transcript release factor, Mouse, Clone: 1F7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PTRF.
Brand: Abnova H00284119-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: KKAFSKEKMEKTKVRTRENLEKTRLKTKENLEKTRHTLEKRMNKLGTRLVPAERREKLKTSRDKLRKSFTPDHVVYARSKTAVYKVPPFSpecifications
| polymerase I and transcript release factor | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_012232 | |
| PTRF | |
| PTRF (NP_036364, 233 a.a. ∼ 321 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (PFA fixed) | |
| 1F7 | |
| Mouse monoclonal antibody raised against a partial recombinant PTRF. | |
| PTRF | |
| FKSG13 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 284119 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction