missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POTEB Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA523543

Product Code. 13248678

  • 520.00 EUR / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

POTEB Polyclonal Antibody for Western Blot, IHC (P), Flow

POTEB (POTE Ankyrin Domain Family Member B) is a Protein Coding gene. An important paralog of this gene is POTEB2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

POTEB
Polyclonal
Unconjugated
POTEB
A26B1; ANKRD26-like family B member 1; ANKRD26-like family B, member 1; cancer/testis antigen family 104, member 5; CT104.5; POTE ankyrin domain family member B; POTE ankyrin domain family member B2; POTE ankyrin domain family member B-like; POTE ankyrin domain family, member B; POTE15; POTE-15; POTEB; POTEB2; Prostate, ovary, testis-expressed protein on chromosome 15; protein expressed in prostate, ovary, testis, and placenta 15
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
100996331
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
A0A0A6YYL3
POTEB
KLH conjugated synthetic peptide between 381-410 amino acids from the C-terminal region of human POTEB.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.