missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Cdc25C (Ser216) Monoclonal Antibody (K.827.0)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA515146
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
M-phase inducer phosphatase 3is an enzyme that in humans is encoded by theCDC25Cgene. This gene is highly conserved during evolution and it plays a key role in the regulation of cell division. The encoded protein is a tyrosine phosphatase and belongs to the Cdc25 phosphatase family. It directs dephosphorylation of cyclin B-bound CDC2 (CDK1) and triggers entry into mitosis. Also, it is thought to suppress p53-induced growth arrest. Multiple alternatively spliced transcript variants of this gene have been described, however, the full-length nature of many of them is not known.
Specifications
| Phospho-Cdc25C (Ser216) | |
| Monoclonal | |
| 170 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| P30307 | |
| Cdc25c | |
| Synthetic phosphopeptide corresponding to residues surrounding pSer216 of human cdc25C. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K.827.0 | |
| Unconjugated | |
| Cdc25c | |
| CDC25; CDC25C; Cdc25m1; cell division cycle 25C; dual specificity phosphatase CDC25C; EC 3.1.3.48; mitosis inducer CDC25; M-phase inducer phosphatase 3; MPIP3; OTTHUMP00000159490; OTTHUMP00000224012; OTTHUMP00000224013; OTTHUMP00000224016; phosphotyrosine phosphatase; PPP1R60; protein phosphatase 1, regulatory subunit 60 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 995 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction