Learn More
p16INK4 with control Mouse anti-Human, Unlabeled, Clone: G175-405, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551154
This item is not returnable.
View return policy
Description
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Specifications
| p16INK4 with control | |
| Monoclonal | |
| Unconjugated | |
| Affinity Purified | |
| RUO | |
| Purified Mouse Anti-Human p16 (150μg) 3 vials of 50μg; HeLa Control Lysate (50μg) | |
| IgG1 |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| G175-405 | |
| Mouse | |
| 150 μg | |
| Primary | |
| Human |
Product Content Correction
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction