Learn More
p16INK4 with control Mouse anti-Human, Unlabeled, Clone: G175-405, BD
Mouse Monoclonal Antibody
Brand:  BD Biosciences 551153
 View more versions of this product
Product Code. 15856558
  • ONLINE EXCLUSIVE PRICE
    183.00 EUR
    182.49 EUR / Each
    Save 0.51 EUR
    0.28% Off
Description
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
TRUSTED_SUSTAINABILITY
Specifications
Specifications
p16INK4 with control
Monoclonal
Unconjugated
Affinity Purified
RUO
Purified Mouse Anti-Human p16 (50μg); HeLa Control Lysate (50μg)
IgG1
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
G175-405
Mouse
50 μg
Primary
Human
Certificates
Promotions
Promotions
For Research Use Only.