missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDLR Mouse anti-Human, Rat, Porcine, Bovine, Clone: C7, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 19 publications
Brand: Novus Biologicals NBP1-78159SS
This item is not returnable.
View return policy
Description
LDLR Monoclonal specifically detects LDLR in Human, Rat, Porcine, Bovine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Radioimmunoassay, Proximity Ligation Assay, Electron Microscopy.Specifications
| LDLR | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry, ELISA, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen, In vitro assay, Radioimmunoassay, Proximity Ligation Assay, Electron Microscopy | |
| P01130, P01131 | |
| LDLR | |
| Partially purified bovine LDL receptor [Swiss-Prot# P01131]. | |
| 0.025 mL | |
| Primary | |
| Human, Rat, Porcine, Bovine | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay, Radioimmune Assays, In situ PLA, Immunoassay | |
| C7 | |
| Unconjugated | |
| Tris-Glycine, 0.15M NaCl with 0.05% Sodium Azide | |
| FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 3949 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only