missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Mouse Endoglin/CD105 (MJ7/18), Proteintech
Rat Monoclonal Antibody
Brand: Proteintech 65048-1-IG-500UG
This item is not returnable.
View return policy
Description
The Endoglin/CD105 antibody from Proteintech is a rat monoclonal antibody generated with inflamed mouse skin of mouse Endoglin/CD105. This antibody recognizes mouse antigen. The Endoglin/CD105 antibody has been validated for the following applications: IF-P, FC analysis.Specifications
| Endoglin/CD105 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| Affinity purification | |
| RUO | |
| 13805 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence, Flow Cytometry | |
| MJ7/18 | |
| Unconjugated | |
| Inflamed mouse skin | |
| 500 μg | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction