Learn More
Invitrogen™ LAP1 Monoclonal Antibody (RL13)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA1074
Description
MA1-074 detects lamin associated polypeptide (LAP) 1A, 1B & 1C from rat, human, and mouse samples. This antibody does not cross-react with LAP 2. MA1-074 has been successfully used in Western blot, immunofluorescence, immunohistochemistry, and immunoprecipitation procedures. By Western blot, this antibody detects 3 distinct proteins of molecular weights ~65, ~56 and ~ 51 kDa representing LAP 1A, 1B, and 1C, respectively in rat liver nuclear envelope fractions. Immunofluorescence staining of LAP 1 in rat liver with MA1-074 results in exclusive labeling of the nuclear periphery and exhibits co-localization with lamin staining. The MA1-074 immunogen is pore complex-lamina fraction isolated from rat liver nuclear envelopes.
Exopeptidase which selectively removes arginine and/or lysine residues from the N-terminus of several peptide substrates including Arg(0)-Leu-enkephalin, Arg(0)-Met-enkephalin and Arg(-1)-Lys(0)-somatostatin-14. Can hydrolyze leukotriene A4 (LTA-4) into leukotriene B4 (LTB-4).
Specifications
| LAP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q5JTV8, Q5PQX1, Q921T2 | |
| TOR1AIP1 | |
| Pore complex-lamina fraction isolated from rat liver nuclear envelopes. | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| RL13 | |
| Unconjugated | |
| TOR1AIP1 | |
| lamina-associated polypeptide 1B; Lamina-associated polypeptide 1C; Lamin-associated protein 1B; LAP1; LAP1B; Lap1c; Tor1aip1; torsin 1A interacting protein 1; torsin A interacting protein 1; torsin-1A-interacting protein 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 208263, 246314, 26092 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.