Learn More
IL-5 Rat anti-Human, Biotin, Clone: JES1-5A10, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554491
Description
The JES1-5A10 antibody reacts with human interleukin-5 (IL-5). The immunogen used to generate the JES1-5A10 hybridoma was recombinant human IL-5. The JES1-5A10 antibody has been reported to cross react with IL-5 from rhesus monkey. This is a neutralizing antibody.
Host Species: Rat
Clone: JES1-5A10
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Recombinant Human IL-5
ELISA
Specifications
| IL-5 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Human IL-5 | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG2a |
| ELISA | |
| JES1-5A10 | |
| Biotin | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.