missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IL-22, PE, Clone: 142928, R&D Systems™

Mouse Monoclonal Antibody has been used in 31 publication
Brand: R&D Systems IC7821P-025
This item is not returnable.
View return policy
Description
IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| IL-22 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 50616 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 142928 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Q9GZX6 | |
| IL22 | |
| E. coli-derived recombinant human IL-22 Ala34-Ile179 Accession # Q9GZX6 | |
| 25 Tests | |
| Primary | |
| Detects human IL-22 in direct ELISAs. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant mouse IL-22 and recombinant rat IL-22 and no cross-reactivity with recombinant human IL-10. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?