Learn More
intercellular adhesion molecule 4 (Landsteiner-Wiener blood group), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ICAM4 protein.
Brand: Abnova H00003386-B01P.50ug
Description
This gene encodes the Landsteiner-Wiener (LW) blood group antigen(s) that belongs to the immunoglobulin (Ig) superfamily, and that shares similarity with the intercellular adhesion molecule (ICAM) protein family. This ICAM protein contains 2 Ig-like C2-type domains and binds to the leukocyte adhesion LFA-1 protein. The molecular basis of the LW(A)/LW(B) blood group antigens is a single aa variation at position 100; Gln-100=LW(A) and Arg-100=LW(B). Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]
Sequence: MGSLFPLSLLFFLAAAYPGVGSALGRRTKRAQSPKGSPLAPSGTSVPFWVRMSPEFVAVQPGKSVQLNCSNSCPQPQNSSLRTPLRQGKTLRGPGWVSYQLLDVRAWSSLAHCLVTCAGKTRWATSRITAYSVPGGLLGGDPEAWKPGHLFRKPGALHRPGSGQRDLDLRVCCWTPRLLAARDLPRAPQSRRPGGPQQLGTHYTDARLEPRAHSFGLRFHRCPCRDPPHCGRCVPMQVPSYEVPGVKGDVLCRLSEKKRNMKQSGEMAIHGGSpecifications
| intercellular adhesion molecule 4 (Landsteiner-Wiener blood group) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ICAM4 protein. | |
| ICAM4 | |
| CD242/LW | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 3386 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001039132.1 | |
| ICAM4 | |
| ICAM4 (NP_001034221.1, 1 a.a. ∼ 272 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.