missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HPV Type 16 E2 Monoclonal Antibody (TVG 261)
GREENER_CHOICE

Product Code. 11580942
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11580942 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11580942 Supplier Invitrogen™ Supplier No. MA190531

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-90531 detects HPV16 E2 from virus samples.

The human papilloma virus (HPV) family of DNA tumor viruses includes HPV-16 and HPV-18, which are associated with a large proportion of cervical cancer cases. E1 and E2 are proteins involved in the regulation of viral DNA replication. Antibodies to this target are useful for detection of protein in cervical lesions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPV Type 16 E2
Applications ELISA, Western Blot
Classification Monoclonal
Clone TVG 261
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias HPV E2; HPV16; HPV-16; human papillomavirus
Host Species Mouse
Immunogen Vaccinia-E2 followed by intravenous injection of the maltose binding protein MBP-E2 (generated from an expression vector in which the E2 ORF was cloned in frame with malE).
Purification Method IgG fraction
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.