Learn More
Anti-Human HLA-DR (L243), Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 65218-1-IG-500UG
 View more versions of this product
Product Code. 30235344
  • ONLINE EXCLUSIVE PRICE
    115.00 EUR
    107.00 EUR / 500µg
    Save 8.00 EUR
    6% Off
Description
The HLA-DR antibody from Proteintech is a mouse monoclonal antibody generated with human lymphoblastoid b-cell line rpmi 8866 of human HLA-DR. This antibody recognizes human antigen. The HLA-DR antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
HLA-DR
Monoclonal
500 μg/mL
HLA-DRA1
Human lymphoblastoid B-cell line RPMI 8866
500 μg
Primary
Human
Flow Antibody
IgG2a κ
Flow Cytometry
L243
Unconjugated
Mouse
Protein A purification
RUO
3122
Store at 2-8°C. Stable for one year after shipment.
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions