Learn More
Anti-Human CD4 (SK3), Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 65147-1-IG-100UG
Product Code. 30234320
  • ONLINE EXCLUSIVE PRICE
    31.21 EUR
    31.00 EUR / 100µg
    Save 0.21 EUR
    0.67% Off
Description
The CD4 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD4. This antibody recognizes human antigen. The CD4 antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
CD4
Monoclonal
500 μg/mL
CD4mut
Affinity purification
RUO
920
Store at 2-8°C. Stable for one year after shipment.
Liquid
Flow Cytometry
SK3
Unconjugated
Mouse
100 μg
Primary
Human
Flow Antibody
IgG1 κ