Learn More
Anti-Human CD4 (OKT4), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65134-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD4 antibody from Proteintech is a mouse monoclonal antibody generated with human peripheral t cells of human CD4. This antibody recognizes human antigen. The CD4 antibody has been validated for the following applications: FC analysis.Specifications
| CD4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| CD4mut | |
| Human peripheral T cells | |
| 500 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG2b κ |
| Flow Cytometry | |
| OKT4 | |
| Unconjugated | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 920 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |