Learn More
HIV2 gp36 Mouse anti-Virus, Clone: 3B10/G2, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17589
Description
MA1-7589 detects HIV from virus samples. MA1-7589 has been successfully used in ELISA, Immunoprecipitation and Western blot procedures. The MA1-7589 immunogen is a purified recombit HIV2 gp36 protein. Store at 4°C short term. For long term storage, aliquot and freeze at -80°C.
Specifications
| HIV2 gp36 | |
| Monoclonal | |
| 1 mg/mL | |
| Human immunodeficiency virus; HIV-2; HIV-2 gp36 | |
| Purified recombit HIV2 gp36 protein. | |
| 100 μg | |
| Primary | |
| 4°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3B10/G2 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.