missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLUT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA511567
This item is not returnable.
View return policy
Description
GLUT2 Polyclonal Antibody for Western Blot, Flow
Glucose transporter 2 isoform is an integral plasma membrane glycoprotein of the liver, islet beta cells, intestine, and kidney epithelium. It mediates facilitated bidirectional glucose transport. Because of its low affinity for glucose, it has been suggested as a glucose sensor. Mutations in the gene that encode this protein cause Fanconi-Bickel syndrome.
Specifications
| GLUT2 | |
| Polyclonal | |
| Unconjugated | |
| Slc2a2 | |
| AI266973; glucose transporter 2; glucose transporter type 2, liver; GLUT2; GLUT-2; GTT2; liver-type glucose transporter; Slc2a2; solute carrier family 2 (facilitated glucose transporter), member 2; solute carrier family 2 member 2; solute carrier family 2, facilitated glucose transporter member 2 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 6514 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 1.8 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P11168 | |
| Slc2a2 | |
| KLH conjugated synthetic peptide between 31-60 amino acids from the N-terminal region of human GLUT2 (SLC2A2). | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction