Learn More
forkhead box A2, Mouse, Clone: 1C7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FOXA2.
Brand: Abnova H00003170-M10.100ug
Description
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific genes such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. This gene has been linked to sporadic cases of maturity-onset diabetes of the young. Transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq
Sequence: HLKPEHHYAFNHPFSINNLMSSEQQHHHSHHHHQPHKMDLKAYEQVMHYPGYGSPMPGSLAMGPVTNKTGLDASPLAADTSYYQGVYSRPIMNSSSpecifications
| forkhead box A2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_021784 | |
| FOXA2 | |
| FOXA2 (NP_068556, 363 a.a. ∼ 457 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| 1C7 | |
| Mouse monoclonal antibody raised against a partial recombinant FOXA2. | |
| FOXA2 | |
| HNF3B/MGC19807/TCF3B | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 3170 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.