missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FICD Polyclonal Antibody

Product Code. 15545792
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15545792 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15545792 Supplier Invitrogen™ Supplier No. PA542933

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: GDVRPFIRFI AKCTETTLDT LLFATTEYSV ALPEAQPNHS GFKETLPVKP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.

Protein that can both mediate the addition of adenosine 5'-monophosphate (AMP) to specific residues of target proteins (AMPylation), and the removal of the same modification from target proteins (de-AMPylation), depending on the context. The side chain of Glu-231 determines which of the two opposing activities (AMPylase or de-AMPylase) will take place. Acts as a key regulator of the ERN1/IRE1-mediated unfolded protein response (UPR) by mediating AMPylation or de-AMPylation of HSPA5/BiP (PubMed:25601083). In unstressed cells, acts as an adenylyltransferase by mediating AMPylation of HSPA5/BiP at 'Thr-518', thereby inactivating it. In response to endoplasmic reticulum stress, acts as a phosphodiesterase by mediating removal of ATP (de-AMPylation) from HSPA5/BiP at 'Thr-518', leading to restore HSPA5/BiP activity. Although it is able to AMPylate RhoA, Rac and Cdc42 Rho GTPases in vitro, Rho GTPases do not constitute physiological substrates (PubMed:19362538, PubMed:25601083). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FICD
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ficd
Gene Accession No. Q9BVA6
Gene Alias adenosine monophosphate-protein transferase FICD; AMPylator FICD; D5Ertd40e; De-AMPylase FICD; FIC domain containing; FIC domain protein adenylyltransferase; FIC domain-containing protein; fic S-phase protein cell division homolog; FICD; HIP13; HIP-13; huntingtin interacting protein 13; huntingtin interacting protein E; huntingtin interactor protein E; huntingtin yeast partner E; Huntingtin-interacting protein 13; huntingtin-interacting protein E; Hype; MGC5623; Protein adenylyltransferase FICD; protein adenylyltransferase FICD; adenosine monophosphate-protein transferase FICD; UNQ3041; UNQ3041/PRO9857
Gene Symbols Ficd
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human FICD (aa 409-458).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11153
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.