Learn More
eukaryotic translation elongation factor 1 gamma, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human EEF1G protein.
Brand: Abnova H00001937-D01P.100ug
Description
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit contains an N-terminal glutathione transferase domain, which may be involved in regulating the assembly of multisubunit complexes containing this elongation factor and aminoacyl-tRNA synthetases. [provided by RefSeq
Sequence: MAAGTLYTYPENWRAFKALIAAQYSGAQVRVLSAPPHFHFGQTNRTPEFLRKFPAGKVPAFEGDDGFCVFESNAIAYYVSNEELRGSTPEAAAQVVQWVSFADSDIVPPASTWVFPTLGIMHHNKQATENAKEEVRRILGLLDAYLKTRTFLVGERVTLADITVVCTLLWLYKQVLEPSFRQAFPNTNRWFLTCINQPQFRAVLGEVKLCEKMAQFDAKKFAETQPKKDTPRKEKGSREEKQKPQAERKEEKKAAAPAPEEEMDECEQALAAEPKAKDPFAHLPKSTFVLDEFKRKYSNEDTLSVALPYFWEHFDKDGWSLWYSEYRFPEELTQTFMSCNLITGMFQRLDKLRKNAFASVILFGTNNSSSISGVWVFRGQELAFPLSPDWQVDYESYTWRKLDPGSEETQTLVREYFSWEGAFQHVGKAFNQGKIFKSpecifications
| eukaryotic translation elongation factor 1 gamma | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human EEF1G protein. | |
| EEF1G | |
| EF1G/GIG35 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 1937 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001404 | |
| EEF1G | |
| EEF1G (NP_001395.1, 1 a.a. ∼ 437 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.