Learn More
DUSP5 (M04), Mouse anti-Human, Clone: 2F3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant DUSP5.
Brand: Abnova H00001847-M04.100ug
Description
The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which are associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates ERK1, is expressed in a variety of tissues with the highest levels in pancreas and brain, and is localized in the nucleus. [provided by RefSeq
Sequence: LKEAFDYIKQRRSMVSPNFGFMGQLLQYESEILPSTPNPQPPSCQGEAAGSSLIGHLQTLSPDMQGAYCTFPASVLAPVPTHSTVSELSRSPVATATSCSpecifications
| DUSP5 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004419 | |
| DUSP5 | |
| DUSP5 (NP_004410, 286 a.a. ∼ 384 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2b κ |
| ELISA, Immunofluorescence, Western Blot | |
| 2F3 | |
| Mouse monoclonal antibody raised against a partial recombinant DUSP5. | |
| DUSP5 | |
| DUSP/HVH3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1847 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.