Learn More
CRX, Mouse, Clone: 2F12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CRX.
Brand: Abnova H00001406-M03.100ug
Description
The protein encoded by this gene is a photoreceptor-specific transcription factor which plays a role in the differentiation of photoreceptor cells. This homeodomain protein is necessary for the maintenance of normal cone and rod function. Mutations in this gene are associated with photoreceptor degeneration, Leber congenital amaurosis type III and the autosomal dominant cone-rod dystrophy 2. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined. [provided by RefSeq
Sequence: MMAYMNPGPHYSVNALALSGPSVDLMHQAVPYPSAPRKQRRERTTFTRSQLEELEALFAKTQYPDVYAREEVALKINLPESRVQVWFKNRRAKCRSpecifications
| CRX | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_000554 | |
| CRX | |
| CRX (NP_000545, 1 a.a. ∼ 95 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2F12 | |
| Mouse monoclonal antibody raised against a partial recombinant CRX. | |
| CRX | |
| CORD2/CRD/LCA7/OTX3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1406 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.