Learn More
Chondroitin-6-sulfate Mouse anti-Chemical, Clone: MK-302, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA183770
Description
Chondroitinase ABC digestion of the target sample is required for antibody activity. Papain or pronase digestion of the target significantly reduces antibody activity. Mouse anti Human chondroitin-6-sulphate antibody, clone MK-302 recognizes chondroitin-6-sulphate, recognizing the chondroitin-6-sulphate stubs of all high density cartilage proteoglycans across all species tested.
This compound belongs to the family of N-acyl-alpha-hexosamines. These are carbohydrate derivatives containing a hexose moeity in which the oxygen atom is replaced by an n-acyl group. [National Center for Biotechnology Information. PubChem Compound Database; CID=5289009]
Specifications
| Chondroitin-6-sulfate | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| 100 μL | |
| Primary | |
| Antibody | |
| IgG1 |
| ELISA | |
| MK-302 | |
| Chondroitin 6′-sulfate; NG6-(3-1)GCU; AC1NRCIM; CHEBI:18296; Chondroitin 6-(hydrogen sulfate) | |
| Chondroitinase ABC-digested adult human aggrecan. | |
| RUO | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.