missing translation for 'onlineSavingsMsg'
Learn More

anti-CD31/PECAM-1 APC, Clone: WM59, Invitrogen™

CD31/PECAM-1 Monoclonal antibody A16224 from Invitrogen specifically detects CD31/PECAM-1 in Human samples. It is validated for Flow Cytometry.

Brand:  Invitrogen A16224

Product Code. 15295974

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

CD31/PECAM-1 Monoclonal antibody A16224 from Invitrogen specifically detects CD31/PECAM-1 in Human samples. It is validated for Flow Cytometry.

CD31 antibodies are a superior marker for angiogenesis, when compared to Factor VIII and CD34 antibodies. CD31 has been used to measure angiogenesis in association with tumor recurrence. CD31 used in a panel with CD34 and Factor VIII has also been used to
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG1, κ
Clone: WM59
Immunogen: human CD31
Formulation: 25 Tests Purified Antibody (0.1mg/ml) in proprietary buffer, with 0.09% sodium azide
Synonyms: CD31 antigen, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, CD31/EndoCAM, adhesion molecule, endoCAM, platelet endothelial cell adhesion molecule
Uniprot Accession: P16284
Entrez GeneID: 5175
Gene Symbol: PECAM1
This product detects CD31/PECAM-1 in Human samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is human CD31.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD31/PECAM-1
Monoclonal
APC
P16284
PECAM1
human CD31
Primary
Human
Purified
Flow Cytometry
WM59
25 Tests Purified Antibody (0.1mg/ml) in proprietary buffer, with 0.09% sodium azide
CD31 antigen, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, CD31/EndoCAM, adhesion molecule, endoCAM, platelet endothelial cell adhesion molecule
Mouse
25 Tests
5175
Store at 4° C
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.