Learn More
CD24 Mouse anti-Human, Brilliant Violet 711, Clone: ML5, BD
Mouse Monoclonal Antibody
205.16 EUR - 486.80 EUR
Specifications
| Antigen | CD24 |
|---|---|
| Clone | ML5 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 711 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15882729
|
BD Biosciences
563371 |
25 Tests |
206.00 EUR 205.16 EUR / Each Save 0.84 EUR 0.41% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15832759
|
BD Biosciences
563401 |
100 Tests |
491.00 EUR 486.80 EUR / Each Save 4.20 EUR 0.86% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ML5 monoclonal antibody specifically binds to CD24. CD24 is a 35-45 kDa two-chain glycoprotein expressed on the surface of B cells, granulocytes and most B-cell lines. CD24 may play a role in the regulation of B-cell proliferation and maturation.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon™ BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy™5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Specifications
| CD24 | |
| Flow Cytometry | |
| Brilliant Violet 711 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| ML5 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG2a κ | |
| Affinity Purified | |
| Heat Stable Antigen Homologue (HSA); Ba-1; CD24A |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.