Learn More
CD18 Mouse anti-Human, PE, Clone: 6.7, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555924
Description
The 6.7 monoclonal antibody specifically binds to β2 integrin, a 90-95 kDa transmembrane protein, which associates non-covalently as a heterodimer with the a chains of CD11a, CD11b, or CD11c. CD18 is expressed on lymphocytes, monocytes, and more weakly on granulocytes. LFA-1 has been shown to play an important role in homotypic and heterotypic cellular adhesion in immune and inflammatory responses.
Specifications
| CD18 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Cloned Human DS6 TCR γδ+ Cells | |
| 100 Tests | |
| Immunology | |
| Cynomolgus, Human, Pig, Rabbit, Rhesus Monkey | |
| IgG1 κ |
| Flow Cytometry | |
| 6.7 | |
| ITGB2; Integrin beta-2; Beta subunit for LFA-1, MAC-1/CR3, and p150,95 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.