missing translation for 'onlineSavingsMsg'
Learn More

Bcl-2 with control Mouse anti-Human, Unlabeled, Clone: 4D7, BD

Product Code. 15806528
Change view
Click to view available options
Quantity:
150 μg
50 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15806528 50 μg Each
15816528 150 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15806528 Supplier BD Biosciences Supplier No. 551097

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The bcl-2 gene was first discovered in t(14:18) follicular lymphomas. Translocation of bcl-2 sequences from chromosome 18 onto the transcriptionally active immunoglobulin locus at chromosome band 14q32 in B-cells deregulates bcl-2 gene expression, resulting in high levels of bcl-2 mRNA. Bcl-2 protein blocks apoptosis (programmed cell death), and thereby may contribute to tomorigenesis by prolonging cell survival rather than by accelerating the rate of cell proliferation. Abundant evidence implicates this protein in the suppression of apoptosis in many types of cells. Bcl-2 is an intracellular membrane protein and resides primarily in the nuclear envelope, outer mitochondrial membrane and endoplasmic reticulum. The reduced molecular weight of Bcl-2 is 26 kDa. \parClone 4D7 recognizes human Bcl-2. A synthetic peptide containing amino acids 61-76 of the human Bcl-2 protein was used as immunogen. This peptide sequence differs from that of mouse Bcl-2 and hence 4D7 does not cross-react with mouse. 4D7 recognizes Bcl-2 as a 26 kDa band; however, additional higher molecular weight bands have been observed in some cell types. These bands may represent oligomers of Bcl-2, which have been reported to occur with members of the Bcl-2 family during the preparation of nonionic detergent extracts.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-2 with control
Applications Immunoassay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4D7
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Includes Purified Mouse Anti-Human Bcl-2 (50μg), Jurkat Cell Lysate (50μg)
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.