missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANO3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559652
This item is not returnable.
View return policy
Description
Immunogen sequence: DKRNTFEKNL RAEGLMLEKE PAIASPDIMF IKIHIPWDTL CKYAERLNIR MPFRKKCYYT DGRSKSMGRM QTYFRRIKNW MAQNPM Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.
The protein encoded by this gene belongs to the TMEM16 family of predicted membrane proteins, that are also known as anoctamins. While little is known about the function of this gene, mutations in this gene have been associated with some cases of autosomal dominant craniocervical dystonia. Cells from individuals with a mutation in this gene exhibited abnormalities in endoplasmic reticulum-dependent calcium signaling. Studies in rat show that the rat ortholog of this protein interacts with, and modulates the activity of a sodium-activated potassium channel. Deletion of this gene caused increased pain sensitivity in the rat model system. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| ANO3 | |
| Polyclonal | |
| Unconjugated | |
| ANO3 | |
| AI838058; ANO3; anoctamin 3; anoctamin-3; anoctamin-3; LOW QUALITY PROTEIN: anoctamin-3; B230324K02Rik; C11orf25; DYT23; DYT24; GENX-3947; Tmem16c; transmembrane protein 16C; transmembrane protein 16C (eight membrane-spanning domains) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 63982 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.30 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BYT9 | |
| ANO3 | |
| Recombinant protein corresponding to Human ANO3. Recombinant protein control fragment (Product #RP-97324). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction