missing translation for 'onlineSavingsMsg'
Learn More

Aniline sulfate, 97%

Product Code. 11473017
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473017 2500 g 2500g
11453017 100 g 100g
11463017 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473017 Supplier Thermo Scientific Alfa Aesar Supplier No. A15563.0E
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 542-16-5 | (C6H5NH2)2·H2SO4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-16-5
Molecular Formula (C6H5NH2)2·H2SO4
MDL Number MFCD00013110
Synonym Benzenamine sulfate 2:1

Specifications

Density 1.377
Odor Odorless
Quantity 2500 g
UN Number UN2811
Beilstein 3729533
Solubility Information Soluble in water
Formula Weight 284.34
Percent Purity 98%
Chemical Name or Material Aniline sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.