Learn More
Aniline Point Standard, ASTM D 611 (Nominal Value: 62.30 °C ), Paragon Scientific™
Product Code. 16070030
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Product Code. 16070030
Brand: Paragon Scientific CRMAPKR
Fuels Testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Aniline Point ASTM D611
  • Certified Reference Material for Aniline Point
  • Manufactured in accordance with ASTM / EN or IP test methods
  • Certified via Round Robin
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom
Specifications
UN Number UN 1223
Quantity 250 mL
Nominal values quoted. Certified Values will vary from batch to batch.