Learn More
Analytical Volumetric Solution Hydrochloric Acid, 6.0N, Reagecon™
Product Code. 13165519 Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Packaging:
1L
Unit Size:
Each
Product Code. 13165519
Brand: Reagecon H26001