Learn More
Analtech Glass Thin Layer Chromatography (TLC) Developing Tank
Shop All Cole Parmer Products
Click to view available options
Dimensions (L x W):
100 x 100 mm
200 x 200 mm
Unit Size:
Each
Description
- Tanks are constructed of durable glass and racks are constructed of anodized aluminum.
- Tanks accommodate standard TLC plates.
- The top surface is ground to a precise tolerance for tight sealing to enable tank saturation and optimal plate development.
- Thinline™ tanks accommodate two plates and require less solvent for development.
- Latch-Lid™ units hold lids in place with stainless steel latches.
- Order rack to develop maximum number of plates in a tank.
Specifications
Specifications
| Material | Glass |
| Quantity | 1 Ea. |
| Type | TLC Tank |
| Dimensions (L x W) | 100 x 100 mm |