Learn More
AN3199, MedChemExpress
Product Code. 16460398
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16460398 5 mg 5mg
16427938 10 mg 10mg
16423877 50 mg 50mg
16413706 25 mg 25mg
4 options
Product Code. 16460398
Brand: Medchem Express HY19830
MedChemExpress AN3199 is a PDE4 inhibitor with an IC50 of 94.5 nM. AN3199 can be used for the research of inflammation-associated diseases such as asthma and chronic obstructive pulmonary disease (COPD).
AN3199 is a PDE4 inhibitor with an IC50 of 94.5 nM. AN3199 can be used for the research of inflammation-associated diseases such as asthma and chronic obstructive pulmonary disease (COPD).
Specifications
Chemical Name or Material AN3199
CAS 1187187-10-5
Color White
Purity Grade Notes Research
Molecular Formula C17H18BNO5
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (305.68 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(OC2=CC=C(B(O)OC3)C3=C2)N=C1)OCCCC
Molecular Weight (g/mol) 327.14
Formula Weight 327.14
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Política de privacidad.