missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin EP Impurity I (Cefadroxil EP Impurity A), Mikromol™

Product Code. 17971146
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17971146

Brand: Mikromol MM0232.09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Amoxicillin EP Impurity I (Cefadroxil EP Impurity A)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-alpha-(4-Hydroxy-phenyl)glycine
CAS 22818-40-2
Analyte or Component Names D-alpha-(4-Hydroxy-phenyl)glycine
Shipping Condition Room Temperature
Molecular Formula C8 H9 N O3
Certifications/Compliance ISO 17025
Synonym (2R)-2-Amino-2-(4-hydroxyphenyl)acetic Acid, Amoxicillin Trihydrate Imp. I (EP), Amoxicillin Imp. I (EP), Cefadroxil Monohydrate Imp. A (EP), Cefadroxil Imp. A (EP), Amoxicillin USP Related Compound I, Amoxicillin USP RC I, (2R)-2-Amino-2-(4-hydroxyphenyl)acetic Acid, Cefprozil EP impurity A, BRL-14969, Amoxycillin related, Ph Eur Amoxicillin Impurity I, D(-)phpg, Amoxicillin BRL-2333, D(-)-p-Hydroxyphenylglycine BRL-14969, Amoxicillin-alpha-penicilloic acid SB-206888, N-(p-Hydroxyphenylglycyl) amoxicillin SB-207459, Dimer SB-590073, Acetaldehyde Adduct 1 SB-648619, Amoxicillin methyl penicilloate SB-206890, Acetaldehyde Adduct 2 SB-292313, Penicilloic Acid Dimer (MW 748 Dimer) SB-647069, Amoxicillin diketopiperazine SB-206891, HPGMe, HPG, HPGMe Test Mix, Amoxicillin trihydrate (Enzymatic) related, Amoxicillin Related Compound I, Amoxicillin Trihydrate Impurity I, Cefadroxil Monohydrate Impurity A, Amoxicillin Impurity I, Cefadroxil Impurity A
InChI Formula InChI=1S/C8H9NO3/c9-7(8(11)12)5-1-3-6(10)4-2-5/h1-4,7,10H,9H2,(H,11,12)/t7-/m1/s1
SMILES N[C@@H](C(=O)O)c1ccc(O)cc1
IUPAC Name (2R)-2-amino-2-(4-hydroxyphenyl)acetic acid
Molecular Weight (g/mol) 167.16
Quantity 100 mg
Formula Weight 167.0582 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.