missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tellurate, 99.5%, Thermo Scientific Chemicals

Product Code. p-7049482
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372438 10 g 10g
11382438 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372438 Supplier Thermo Scientific Alfa Aesar Supplier No. 014487.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-06-0
Molecular Formula H8N2O4Te
Molecular Weight (g/mol) 227.67
MDL Number MFCD00015964
InChI Key DRGYXGZFRXFMHF-UHFFFAOYSA-N
Synonym ammonium tellurate, diammonium tellurate, diammonium tetraoxotellurate, ammonium tellurate nh4 2teo4, telluric acid, diammonium salt, telluric acid h2teo4 , diammonium salt, ammonium tellurate vi nh4 2teo4, telluric acid h2teo4 , ammonium salt 1:2, ammonium tellurate vi 6ci, diazanium tellurate
PubChem CID 61608
IUPAC Name diazanium;tellurate
SMILES [NH4+].[NH4+].[O-][Te]([O-])(=O)=O

Specifications

Color White
Physical Form Powder
Quantity 10 g
UN Number UN3284
Solubility Information Soluble in water.
Formula Weight 227.67
Percent Purity 99.5%
Chemical Name or Material Ammonium tellurate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.