missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfamate, 98+%

Product Code. p-7023794
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453278 250 g 250g
11443278 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11453278 Supplier Thermo Scientific Alfa Aesar Supplier No. A17696.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7773-06-0 | H6N2O3S | 114.119 g/mol

Ammonium sulfamate is applied in water solution or oil-water emulsion as a leaf, or foliar spray for control of woody plants, or it is applied as crystals or concentrated solution to cuts in the bark or on freshly-cut stumps of undesirable trees to prevent resprouting. Used as a compost accelerator, flame retardant and in industrial processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium sulfamate is applied in water solution or oil-water emulsion as a leaf, or foliar spray for control of woody plants, or it is applied as crystals or concentrated solution to cuts in the bark or on freshly-cut stumps of undesirable trees to prevent resprouting. Used as a compost accelerator, flame retardant and in industrial processes.

Solubility
Soluble in water(2800g/L ). Insoluble in methanol, ether, n-octanol.

Notes
Hygroscopic. Store at room temperature. Stored in airtight containers which are kept closed and away from air and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7773-06-0
Molecular Formula H6N2O3S
Molecular Weight (g/mol) 114.119
MDL Number MFCD00011429
InChI Key GEHMBYLTCISYNY-UHFFFAOYSA-N
Synonym ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
PubChem CID 24482
ChEBI CHEBI:81950
IUPAC Name azanium;sulfamate
SMILES [NH4+].NS(=O)(=O)[O-]

Specifications

Boiling Point 160°C (decomposition)
Melting Point 131°C to 135°C
pH 5.2
Quantity 250 g
Linear Formula NH4SO3NH2
Merck Index 14,554
Solubility Information Soluble in water(2800g/L ). Insoluble in methanol,ether,n-octanol.
Formula Weight 114.12
Percent Purity ≥98%
Odor Weak
Sensitivity Hygroscopic
Chemical Name or Material Ammonium sulfamate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium sulfamate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium sulfamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.