missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate (para) hydrate, Puratronic™, 99.999% (metals basis excluding W), W 300ppm max

Product Code. p-4407225
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11356927

Brand: Thermo Scientific Alfa Aesar 010811.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2127839-79-4 | H24Mo7N6O24 | 1163.85 g/mol

It is used for ceramics, decorating, electroplating, fixing of metals, fertilizers, photography, and as a negative stain in electron microscopy. It is also used in the manufacture of molybdenum. It is a useful reagent for detecting and determining phosphates, arsenates, lead and some alkaloids. It is useful in the preparation of catalysts for desulfurization and dehydrogenation reactions. Ammonium molybdate has been reported to control polycondensation reactions and decrease shrinkage values of metakaolin-based geopolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2127839-79-4
Molecular Formula H24Mo7N6O24
Molecular Weight (g/mol) 1163.85
MDL Number MFCD00064633
InChI Key MISTUIVWPMGXFX-UHFFFAOYSA-T
Synonym hexaammonium molybdate, ammoniummolybdatehydrate, nh4 6mo7o4
PubChem CID 71306766
IUPAC Name hexaazanium;dioxido(dioxo)molybdenum
SMILES [NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O=[Mo]123(=O)O[Mo]45(=O)(=O)O[Mo]67(=O)(=O)O[Mo]89(=O)(=O)O[Mo]%10%11(=O)(=O)O[Mo]%12(=O)(=O)(O1)[O+]2[Mo]([O+]34)([O+]56)([O+]%10%12)([O+]78)[O+]9%11

Specifications

Melting Point ∽190°C (decomposition)
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Freely soluble in water.
Formula Weight 1163.79 (Anhydrous)
Appearance Powder
Density 2.498 g/cm3
Chemical Name or Material Ammonium molybdate (para) hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.