missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachlororuthenate(IV), Ru 28.4% min

Product Code. p-7048888
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11311958

Brand: Thermo Scientific Alfa Aesar 011874.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18746-63-9 | Cl6H8N2Ru | 349.85 g/mol

Ammonium hexachlororuthenate(IV) is used in laboratory chemicals and manufacture of other substances. Ammonium hexachlororuthenate (ACR) is a salt precipitated during a substitution reaction between ammonium and ruthenate dissolved in a strong hydrochloric acid medium. This precipitation reaction is used within the platinum industry as a means of recovering and purifying ruthenium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18746-63-9
Molecular Formula Cl6H8N2Ru
Molecular Weight (g/mol) 349.85
MDL Number MFCD00015962
InChI Key DHTSBQSPUBOKKI-UHFFFAOYSA-J
Synonym ammonium hexachlororuthenate iv, diammonium hexachlororuthenate, ammonium chlororuthenate, ammonium hexachlororuthenate, epitope id:151520, ruthenium iv-ammonium chloride, ammonium hexachloridoruthenate iv, ammonium hexachloridoruthenate 2-, diammonium hexachlororuthenate 2-
PubChem CID 46173598
ChEBI CHEBI:60134
SMILES [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+4]

Specifications

Color Red to Brown
Physical Form Crystalline Powder
Quantity 1 g
Solubility Information Soluble in water.
IUPAC Name diazanium;hexachlororuthenium(2-)
Formula Weight 349.87
Concentration or Composition (by Analyte or Components) Ru 28.4% min.
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Ammonium hexachlororuthenate(IV)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.