missing translation for 'onlineSavingsMsg'
Learn More

Ammonium-d{4} deuteroxide, 99% (Isotopic), 23.0-27.0 wt% soln. in D{2}O

Product Code. p-7049810
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302598 1 g 1g
11312598 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11302598 Supplier Thermo Scientific Alfa Aesar Supplier No. 022980.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 12168-30-8 | H5NO | 40.077 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12168-30-8
Molecular Formula H5NO
Molecular Weight (g/mol) 40.077
MDL Number MFCD00064511
InChI Key VHUUQVKOLVNVRT-NSPFYZSMSA-N
Synonym ammonium deuteroxide, ammonium-d4 deuteroxide solution, 25 wt. % in d2o, 99 atom % d
PubChem CID 16211396
SMILES N.O

Specifications

Physical Form Liquid
Color Colorless
Refractive Index 1.334
Sensitivity Moisture
Quantity 1 g
Linear Formula ND4OD
UN Number UN2672
Formula Weight 40.09
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 23.0 to 27.0 wt% soln. in D2O
Chemical Name or Material Ammonium-d4 deuteroxide, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.