missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate-d7, TRC

Product Code. 30337957
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30337957 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30337957 Supplier Toronto Research Chemicals Supplier No. TRCA633901500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium Acetate-d7
CAS 194787-05-8
Molecular Formula C2 D3 O2 . D4 N
Quantity 500 mg
Synonym Acetic-d3 acid-d, ammonium-d4 salt (9CI), Ammonium-d4 acetate-d3, Ammonium acetate-d7, Ammonium acetate D7
InChI Formula InChI=1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3/i1D3;/hD4
SMILES [2H]C([2H])([2H])C(=O)[O-].[2H][N+]([2H])([2H])[2H]
IUPAC Name tetradeuterioazanium;2,2,2-trideuterioacetate
Molecular Weight (g/mol) 84.126
Formula Weight 84.092
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.