Learn More
Ammonia solution 25%, BAKER ANALYZED™, J.T. Baker™, Avantor™
Product Code. 10700312 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10700312 1 L 1 L
10691552 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10700312 Supplier JT Baker Supplier No. 6051.1000
Laboratory Reagent
Chemical Identifiers
CAS 1336-21-6
Molecular Formula NH3
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
Synonym Ammonium hydroxide
Specifications
Boiling Point 32°C to 37.7°C (1013 hPa)
Melting Point -60°C to -57.5°C
Density 0.88 - 0.91 g/cm³ (20°C)
Quantity 1 L
Packaging Glass bottle
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.